Bis(hexafluoroacetylacetonato)manganese(II) hydrate ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

H0557-1G H0557-5G
TCIAH0557-1GEA 42 EUR
TCIAH0557-1G TCIAH0557-5G
Bis(hexafluoroacetylacetonato)manganese(II) hydrate ≥95.0% (by titrimetric analysis)
Bis(hexafluoroacetylacetonato)manganese(II) hydrate
Formula: C₁₀H₂F₁₂MnO₄
Número MDL: MFCD00042513
Núm. CAS: 19648-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2197bb3c9048d49c02937818a4239442f94fa1f5&vid=c6d631ef51a89863703fe07b2ad40958588d2df4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 95.0 % (calcd.on anh.substance)
Water max. 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR