2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(2-Benzotriazolyl)-p-cresol, Drometrizole

H0560-25G H0560-500G
TCIAH0560-25GEA 16 EUR
TCIAH0560-25G TCIAH0560-500G
2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99.0% (by HPLC, titration analysis)
2-(2-Hydroxy-5-methylphenyl)benzotriazole
Formula: C₁₃H₁₁N₃O
MW: 225,25 g/mol
Número MDL: MFCD00022903
Núm. CAS: 2440-22-4
EINECS: 219-470-5
Índice Merck: 14,03447

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 129.0 to 133.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR