4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine ≥98.0% (by titrimetric analysis) for determination of Co(III), Fe(II)

Proveedor: TCI
Atención

Sinónimos: 6-Amino-2-dimethylamino-5-nitroso-4-pyrimidinol, 4-Hydroxy-2-dimethylamino-5-nitroso-6-aminopyrimidine

H0562-1G H0562-100MG
TCIAH0562-1GEA 298 EUR
TCIAH0562-1G TCIAH0562-100MG
4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine ≥98.0% (by titrimetric analysis) for determination of Co(III), Fe(II)
4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine
Formula: C₆H₉N₅O₂
MW: 183,17 g/mol
Número MDL: MFCD00059767
Núm. CAS: 70700-44-6

Order Now

Resultados test especificaciones

Appearance Red to Dark red to Dark purple powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR