2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Hydroxy-2-methylbutanoic acid

H0636-25G H0636-5G
TCIAH0636-25GEA 119 EUR
TCIAH0636-25G TCIAH0636-5G
2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-2-methylbutyric acid
Formula: C₅H₁₀O₃
MW: 118,13 g/mol
Pto. Ebullición: 109°C
Número MDL: MFCD00004476
Núm. CAS: 3739-30-8
EINECS: 223-120-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 74.0 deg-C
Solubility in Water within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR