Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
N-hexadecyltrimethylammoniumtetrafluoroborate, Cetyltrimethylammonium Tetrafluoroborate
H0734-1G
H0734-5G
TCIAH0734-1GEA
87
EUR
TCIAH0734-1G
TCIAH0734-5G
Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium tetrafluoroborate
Formula:
C₁₉H₄₂BF₄N MW: 371,36 g/mol |
Número MDL:
MFCD00144896 Núm. CAS: 73257-08-6 UN: 1759 ADR: 8,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...