Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: Cetyltrimethylammonium Perchlorate

H0735-1G H0735-5G
TCIAH0735-1GEA 87 EUR
TCIAH0735-1G TCIAH0735-5G
Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium perchlorate
Formula: C₁₉H₄₂ClNO₄
Número MDL: MFCD00144895
Núm. CAS: 6941-37-3
UN: 1479
ADR: 5.1,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Methanol almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR