Methyl-4-(2-hydroxyethoxy)benzoate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Methyl 4-(2-hydroxyethoxy)benzoate, 4-(2-Hydroxyethoxy)benzoic Acid Methyl Ester

H0820-25G H0820-5G
TCIAH0820-25GEA 285 EUR
TCIAH0820-25G TCIAH0820-5G
Methyl-4-(2-hydroxyethoxy)benzoate ≥98.0% (por GC)
Methyl-4-(2-hydroxyethoxy)benzoate
Formula: C₁₀H₁₂O₄
Número MDL: MFCD00075872
Núm. CAS: 3204-73-7
EINECS: 221-708-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR