Methyl (4-hydroxybenzoyl)acetate ≥93.0% (por GC)
Proveedor: TCI
Sinónimos:
Methyl 3-(4-hydroxyphenyl)-3-oxopropanoate, (4-Hydroxybenzoyl)acetic Acid Methyl Ester, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate
H0828-1G
H0828-5G
TCIAH0828-1GEA
149
EUR
TCIAH0828-1G
TCIAH0828-5G
Methyl (4-hydroxybenzoyl)acetate ≥93.0% (por GC)
Methyl (4-hydroxybenzoyl)acetate
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol |
Número MDL:
MFCD00191516 Núm. CAS: 32066-29-8 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 93.0 % (total of isomer) |
Melting point | 85.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...