ɑ,ɑ,ɑ',ɑ',4,5-Hexabromo-o-xylene ≥98.0% (por GC)

Proveedor: TCI
Peligro

H0831-1G H0831-5G
TCIAH0831-1GEA 90 EUR
TCIAH0831-1G TCIAH0831-5G
ɑ,ɑ,ɑ',ɑ',4,5-Hexabromo-o-xylene ≥98.0% (por GC)
alpha,alpha,alpha',alpha',4,5-Hexabromo-o-xylene
Formula: C₈H₄Br₆
MW: 579,54 g/mol
Número MDL: MFCD00191445
Núm. CAS: 13209-20-6
UN: 1759
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 139.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR