2-Hydroxy-3-butynoic acid ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
H0905-1G
H0905-100MG
TCIAH0905-1GEA
868
EUR
TCIAH0905-1G
TCIAH0905-100MG
2-Hydroxy-3-butynoic acid ≥97.0% (by titrimetric analysis)
2-Hydroxy-3-butynoic acid
Formula:
C₄H₄O₃ MW: 100,07 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00671546 Núm. CAS: 38628-65-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...