4-Hydroxymethyl-5-methylimidazole ≥96.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
(5-Methyl-1H-imidazol-4-yl)methanol
H0976-25G
H0976-5G
TCIAH0976-25GEA
174
EUR
TCIAH0976-25G
TCIAH0976-5G
4-Hydroxymethyl-5-methylimidazole ≥96.0% (by GC, titration analysis)
4-Hydroxymethyl-5-methylimidazole
Formula:
C₅H₈N₂O MW: 112,13 g/mol |
Número MDL:
MFCD00792476 Núm. CAS: 29636-87-1 EINECS: 249-740-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 134.0 to 138.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...