D-Homophenylalanine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
(R)-2-Amino-4-phenylbutyric Acid, H-D-HomoPhe-OH, H-D-HoPhe-OH
H0984-1G
H0984-100MG
TCIAH0984-1GEA
131
EUR
TCIAH0984-1G
TCIAH0984-100MG
D-Homophenylalanine ≥98.0% (by titrimetric analysis)
D-Homophenylalanine
Formula:
C₁₀H₁₃NO₂ MW: 179,22 g/mol |
Número MDL:
MFCD00063091 Núm. CAS: 82795-51-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -44.0 to -46.0 deg (C=1, 3mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...