4-(Hydroxymethyl)phenoxymethyl polystyrene resin 1,0 - 2,0 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Proveedor: TCI
H0987-25G H0987-5G
TCIAH0987-25GEA 319 EUR
TCIAH0987-25G TCIAH0987-5G
4-(Hydroxymethyl)phenoxymethyl polystyrene resin 1,0 - 2,0 mmol/g, cross-linked with 1% DVB (200-400 mesh)
4-(Hydroxymethyl)phenoxymethyl polystyrene resin
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87dd5fa98f100cee48198c2d1be44ae0b86ab6f8&vid=9b961e9d3518f9a9c4e654944b24f251adf0a9c5&prid=web&clid=SAPDAM
Order Now
Resultados test especificaciones
Loading 1.00 to 2.00 mmol/g