Hexyldimethyloctylammonium bromide ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-Hexyl-N,N-dimethyloctan-1-aminium bromide
H0989-25G
H0989-5G
TCIAH0989-25GEA
223
EUR
TCIAH0989-25G
TCIAH0989-5G
Hexyldimethyloctylammonium bromide ≥97.0% (by titrimetric analysis)
Hexyldimethyloctylammonium bromide
Formula:
C₁₆H₃₆BrN MW: 322,38 g/mol |
Número MDL:
MFCD03093632 Núm. CAS: 187731-26-6 |
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...