2-Hydroxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
H1016-25G
H1016-5G
TCIAH1016-25GEA
175
EUR
TCIAH1016-25G
TCIAH1016-5G
2-Hydroxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Hydroxyquinoline-4-carboxylic acid
Formula:
C₁₀H₇NO₃ |
Número MDL:
MFCD00023942 Núm. CAS: 15733-89-8 EINECS: 239-827-9 |
|
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...