3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
H1019-1G
H1019-5G
TCIAH1019-1GEA
69
EUR
TCIAH1019-1G
TCIAH1019-5G
3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone
Formula:
C₇H₈O₅ MW: 172,14 g/mol |
Número MDL:
MFCD00110732 Núm. CAS: 2029-49-4 |
Resultados test especificaciones
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point (Decomposition) | 145.0 to 153.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...