4-Hydroxyphenyldimethylsulphonium methyl sulphate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(4-Hydroxyphenyl)dimethylsulphonium methyl sulphate, Dimethyl-4-hydroxyphenylsulphonium Methyl Sulphate
H1022-25G
H1022-5G
TCIAH1022-25GEA
252
EUR
TCIAH1022-25G
TCIAH1022-5G
4-Hydroxyphenyldimethylsulphonium methyl sulphate ≥98.0% (by titrimetric analysis)
4-Hydroxyphenyldimethylsulphonium methyl sulphate
Formula:
C₉H₁₄O₅S₂ MW: 266,33 g/mol |
Número MDL:
MFCD08276380 Núm. CAS: 32279-04-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...