Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetic Acid Methyl Ester

H1049-1G H1049-100MG
TCIAH1049-1GEA 412 EUR
TCIAH1049-1G TCIAH1049-100MG
Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate ≥98.0% (por GC)
Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate
Formula: C₁₀H₁₁NO₆
MW: 241.20 g/mol
Número MDL: MFCD04117920
Núm. CAS: 308815-81-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR