Hydrazine acetate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

H1112-25G H1112-5G
TCIAH1112-25GEA 220 EUR
TCIAH1112-25G TCIAH1112-5G
Hydrazine acetate ≥98.0% (by titrimetric analysis)
Hydrazine acetate
Formula: H₂NNH₂·CH₃CO₂H
MW: 92.098 g/mol
Número MDL: MFCD00013141
Núm. CAS: 7335-65-1
UN: 3263
ADR: 8 II

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR