Hexadecyl lactate ≥75.0% (por GC)

Proveedor: TCI

Sinónimos: Lactic Acid Hexadecyl Ester, Hexadecyl 2-Hydroxypropionate, Cetyl Lactate

H1159-25G H1159-500G
TCIAH1159-25GEA 16 EUR
TCIAH1159-25G TCIAH1159-500G
Hexadecyl lactate ≥75.0% (por GC)
Hexadecyl lactate
Formula: C₁₉H₃₈O₃
MW: 314.51 g/mol
Núm. CAS: 35274-05-6
EINECS: 252-478-7
Índice Merck: 14,02030

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 75.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR