2,3,6,7,10,11-Hexabromotriphenylene ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

H1201-1G H1201-100MG
TCIAH1201-1GEA 416 EUR
TCIAH1201-1G TCIAH1201-100MG
2,3,6,7,10,11-Hexabromotriphenylene ≥98.0% (by titrimetric analysis)
2,3,6,7,10,11-Hexabromotriphenylene
Formula: C₁₈H₆Br₆
MW: 701,67 g/mol
Número MDL: MFCD09834716
Núm. CAS: 82632-80-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR