(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥98.0% (by GC, total nitrogen)
Proveedor: TCI
|
Atención
|
H1241-1G
H1241-5G
TCIAH1241-1GEA
32
EUR
TCIAH1241-1G
TCIAH1241-5G
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥98.0% (by GC, total nitrogen)
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
Formula:
C₅H₉NO₂ MW: 115,13 g/mol |
Número MDL:
MFCD00077791 Núm. CAS: 66673-40-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...