Azetidin-3-ol hydrochloride ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: 3-Hydroxyazetidine hydrochloride

H1264-1G H1264-5G
TCIAH1264-1GEA 39 EUR
TCIAH1264-1G TCIAH1264-5G
Azetidin-3-ol hydrochloride ≥98.0% (by total nitrogen basis)
Azetidin-3-ol hydrochloride
Formula: C₃H₈ClNO
MW: 109,56 g/mol
Número MDL: MFCD02683887
Núm. CAS: 18621-18-6

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 87.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR