8-Hydroxyquinoline-N-Oxide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

H1272-25G H1272-5G
TCIAH1272-25GEA 121 EUR
TCIAH1272-25G TCIAH1272-5G
8-Hydroxyquinoline-N-Oxide ≥98.0% (by HPLC, titration analysis)
8-Hydroxyquinoline-N-Oxide
Formula: C₉H₇NO₂
Número MDL: MFCD00006739
Núm. CAS: 1127-45-3
EINECS: 214-430-3

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR