(R)-2-Hydroxy-4-phenylbutyric acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI

H1338-1G H1338-5G
TCIAH1338-1GEA 69 EUR
TCIAH1338-1G TCIAH1338-5G
(R)-2-Hydroxy-4-phenylbutyric acid ≥97.0% (by GC, titration analysis)
(R)-2-Hydroxy-4-phenylbutyric acid
Formula: C₁₀H₁₂O₃
MW: 180,20 g/mol
Número MDL: MFCD00192219
Núm. CAS: 29678-81-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 114.0 to 118.0 deg-C
Specific rotation [a]20/D -7.0 to -10.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR