Halosulphuron methyl ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulphonyl]-1-methyl-1H-pyrazole-4-carboxylic acid Methyl Ester, Methyl 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulphonyl]-1-methyl-1H-pyrazole-4-carboxylate

H1374-5G H1374-1G
TCIAH1374-5GEA 274 EUR
TCIAH1374-5G TCIAH1374-1G
Halosulphuron methyl ≥98.0% (by HPLC, titration analysis)
Halosulphuron methyl
Formula: C₁₃H₁₅ClN₆O₇S
MW: 434,81 g/mol
Número MDL: MFCD01631160
Núm. CAS: 100784-20-1
UN: 3077
ADR: 9,III
Índice Merck: 14,04602

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR