6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
6-Hydroxy-4-pyrimidinecarboxylic acid, 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic acid
H1392-200MG
H1392-1G
TCIAH1392-200MGEA
28
EUR
TCIAH1392-200MG
TCIAH1392-1G
6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Hydroxypyrimidine-4-carboxylic acid
Formula:
C₅H₄N₂O₃ MW: 140,0969 g/mol |
Número MDL:
MFCD00023267 Núm. CAS: 6299-87-2 |
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...