3-Hydroxy-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Hydroxy-3-phenylpropionic acid
H1394-1G
H1394-5G
TCIAH1394-1GEA
18
EUR
TCIAH1394-1G
TCIAH1394-5G
3-Hydroxy-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-3-phenylpropanoic acid
Formula:
C9H10O3 MW: 166,1700 g/mol |
Número MDL:
MFCD00137907 Núm. CAS: 3480-87-3 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...