2-Hydroxy-2-azaadamantane ≥98.0% (by GC, titration analysis)

Proveedor: TCI

H1404-200MG H1404-1G H1404-5G
TCIAH1404-200MGEA 64 EUR
TCIAH1404-200MG TCIAH1404-1G TCIAH1404-5G
2-Hydroxy-2-azaadamantane ≥98.0% (by GC, titration analysis)
2-Hydroxy-2-azaadamantane
Formula: C₉H₁₅NO
Núm. CAS: 1155843-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a62d99a088da4c0ffa711a32a83b7f494ed4c78&vid=0a62d99a088da4c0ffa711a32a83b7f494ed4c78&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR