(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh's Catalyst, (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide

H1407-200MG H1407-1G
TCIAH1407-200MGEA 77 EUR
TCIAH1407-200MG TCIAH1407-1G
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide ≥98.0% (by HPLC, titration analysis)
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide
Formula: C₂₃H₃₀N₂O₂
MW: 366,51 g/mol
Número MDL: MFCD22200506
Núm. CAS: 910110-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88d0403dfa604574c57a839e55285e556470d8d8&vid=88d0403dfa604574c57a839e55285e556470d8d8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 191.0 to 195.0 deg-C
Specific rotation [a]20/D -46.0 to -49.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR