1-Hexil-3-metilimidazolio bis(trifluorometilsulfonil)imida ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3-Hexyl-1-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide, 1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide

H1423-5G H1423-25G
TCIAH1423-5GEA 61 EUR
TCIAH1423-5G TCIAH1423-25G
1-Hexil-3-metilimidazolio bis(trifluorometilsulfonil)imida ≥98.0% (by HPLC, titration analysis)
1-Hexil-3-metilimidazolio bis(trifluorometilsulfonil)imida
Formula: C₁₂H₁₉F₆N₃O₄S₂
MW: 447.42 g/mol
Pto. de fusión: –9°C
Número MDL: MFCD08458931
Núm. CAS: 382150-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bae4de598aa275c62826d14ca8887ef26601d68f&vid=bae4de598aa275c62826d14ca8887ef26601d68f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR