2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene ≥98.0% (by HPLC)
Proveedor: TCI
H1449-200MG
H1449-1G
TCIAH1449-200MGEA
111
EUR
TCIAH1449-200MG
TCIAH1449-1G
2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene ≥98.0% (by HPLC)
2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene
Formula:
C₅₄H₈₄O₆ MW: 829,26 g/mol |
Número MDL:
MFCD16250223 Núm. CAS: 70351-86-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...