2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene ≥98.0% (by HPLC)
Proveedor: TCI
H1450-200MG
H1450-1G
TCIAH1450-200MGEA
140
EUR
TCIAH1450-200MG
TCIAH1450-1G
2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene ≥98.0% (by HPLC)
2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene
Formula:
C₆₆H₁₀₈O₆ MW: 997,58 g/mol |
Número MDL:
MFCD20528916 Núm. CAS: 70351-87-0 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...