1-Hydroxy-6-(trifluoromethyl)benzotriazole ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

H1454-1G H1454-5G
TCIAH1454-1GEA 48 EUR
TCIAH1454-1G TCIAH1454-5G
1-Hydroxy-6-(trifluoromethyl)benzotriazole ≥98.0% (by HPLC, titration analysis)
1-Hydroxy-6-(trifluoromethyl)benzotriazole
Formula: C₇H₄F₃N₃O
MW: 203,12 g/mol
Número MDL: MFCD00038806
Núm. CAS: 26198-21-0
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b3e84b8110398763458925e520bedcdd84e8cfa&vid=0b3e84b8110398763458925e520bedcdd84e8cfa&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR