2'-Hidroxi-5'-metil-3'-nitroacetofenona ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
H1479-5G
H1479-25G
TCIAH1479-5GEA
40
EUR
TCIAH1479-5G
TCIAH1479-25G
2'-Hidroxi-5'-metil-3'-nitroacetofenona ≥98.0% (by GC, titration analysis)
2'-Hidroxi-5'-metil-3'-nitroacetofenona
Formula:
C₉H₉NO₄ MW: 195,17 g/mol |
Número MDL:
MFCD00192216 Núm. CAS: 66108-30-3 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...