1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2'-Hydroxy-2-(4-methoxybenzal)acetophenone, 2'-Hydroxy-4-methoxychalcone, 1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one, 2'-Hydroxy-2-(4-methoxybenzylidene)acetophenone

H1484-1G H1484-5G
TCIAH1484-1GEA 69 EUR
TCIAH1484-1G TCIAH1484-5G
1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one ≥98.0% (by HPLC, titration analysis)
1-(2-Hydroxyphenyl)-3-(4-methoxyphenyl)-2-propen-1-one
Formula: C₁₆H₁₄O₃
MW: 254,29 g/mol
Número MDL: MFCD00016447
Núm. CAS: 3327-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5d52c96d488d3c47fbb42e149c83d4ad574741&vid=aa5d52c96d488d3c47fbb42e149c83d4ad574741&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR