4-Hexylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Hexylphenylboronic Acid, 4-N-Hexylphenylboronic acid, 4-(Hex-1-yl)benzeneboronic acid
H1489-1G
H1489-5G
TCIAH1489-1GEA
60
EUR
TCIAH1489-1G
TCIAH1489-5G
4-Hexylphenylboronic acid (contains varying amounts of Anhydride)
4-Hexylphenylboronic acid
Formula:
C₁₂H₁₉BO₂ MW: 206.09 g/mol |
Número MDL:
MFCD04039029 Núm. CAS: 105365-50-2 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...