6-Hydroxy-3-coumaranone ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
H1511-1G
H1511-5G
TCIAH1511-1GEA
25
EUR
TCIAH1511-1G
TCIAH1511-5G
6-Hydroxy-3-coumaranone ≥97.0% (by GC, titration analysis)
6-Hydroxy-3-coumaranone
Formula:
C₈H₆O₃ |
Número MDL:
MFCD00068174 Núm. CAS: 6272-26-0 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...