Hidrazina bromhidrato ∼50% in water

Proveedor: TCI
Peligro

Sinónimos: Hydrazine monohydrobromide

H1517-25G H1517-500G
TCIAH1517-25GEA 16 EUR
TCIAH1517-25G TCIAH1517-500G
Hidrazina bromhidrato ∼50% in water
Hidrazina bromhidrato
Formula: H₅BrN₂
MW: 112,96 g/mol
Número MDL: MFCD00065394
Núm. CAS: 13775-80-9
EINECS: 237-412-7
UN: 3289
ADR: 6.1,II

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Concentration (Potassium iodate Method) 49.0 to 51.0 w/w%
Concentration (Argentmetric Titration) 49.0 to 51.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR