Hexafluoroisopropyl methyl ether ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Methyl hexafluoroisopropyl ether
H1524-5G
H1524-25G
TCIAH1524-5GEA
54
EUR
TCIAH1524-5G
TCIAH1524-25G
Hexafluoroisopropyl methyl ether ≥98.0% (por GC)
Hexafluoroisopropyl methyl ether
Formula:
C₄H₄F₆O MW: 182,064 g/mol Pto. Ebullición: 50°C |
Número MDL:
MFCD00221772 Núm. CAS: 13171-18-1 UN: 3271 ADR: 3,II |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...