17alpha-Hydroxyprogesterone Acetate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 17alpha-Hydroxypregn-4-ene-3,20-diene Acetate, 17alpha-Acetoxypregn-4-ene-3,20-dione, 17alpha-Acetoxyprogesterone

H1527-1G H1527-5G
TCIAH1527-1GEA 25 EUR
TCIAH1527-1G TCIAH1527-5G
17alpha-Hydroxyprogesterone Acetate ≥98.0% (by HPLC)
17alpha-Hydroxyprogesterone Acetate
Formula: C₂₃H₃₂O₄
MW: 372.51 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00021152
Núm. CAS: 302-23-8
EINECS: 206-119-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 244.0 to 248.0 deg-C
Specific rotation [a]20/D +71.0 to +76.0 deg (C=1, CHCL3)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR