3-Hydroxy-1-methacryloyloxyadamantane ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Hydroxy-1-adamantyl methacrylate, 3-Hydroxyadamantan-1-yl methacrylate, Methacrylic acid 3-Hydroxy-1-adamantyl ester
H1543-5G
H1543-25G
TCIAH1543-5GEA
82
EUR
TCIAH1543-5G
TCIAH1543-25G
3-Hydroxy-1-methacryloyloxyadamantane ≥98.0% (por GC)
3-Hydroxy-1-methacryloyloxyadamantane
Formula:
C₁₄H₂₀O₃ MW: 236,31 g/mol |
Número MDL:
MFCD13152044 Núm. CAS: 115372-36-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...