2-Hydroxyflutamide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Hydroxy-2-methyl-N-(4-nitro-3-(trifluoromethyl)phenyl)propenamide, Hydroxyflutamide
H1600-25MG
H1600-100MG
TCIAH1600-25MGEA
62
EUR
TCIAH1600-25MG
TCIAH1600-100MG
2-Hydroxyflutamide ≥98.0% (by HPLC)
2-Hydroxyflutamide
Formula:
C₁₁H₁₁F₃N₂O₄ MW: 292,21 g/mol |
Número MDL:
MFCD00563126 Núm. CAS: 52806-53-8 UN: 3077 ADR: 9,III |
Resultados test especificaciones
Appearance | White to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 9.00 to 10.00 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...