Heptadecan-9-amine ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
H1640-1ML
H1640-5ML
TCIAH1640-1MLEA
135
EUR
TCIAH1640-1ML
TCIAH1640-5ML
Heptadecan-9-amine ≥96.0% (by GC, titration analysis)
Heptadecan-9-amine
Formula:
C₁₇H₃₇N Pto. Ebullición: 120°C |
Número MDL:
MFCD30742905 Núm. CAS: 3241-20-1 UN: 2735 ADR: 8,II |
|
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...