L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: LD-Heptose Peracetate

H1657-20MG H1657-100MG
TCIAH1657-20MGEA 59 EUR
TCIAH1657-20MG TCIAH1657-100MG
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate ≥98.0% (por GC)
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate
Formula: C₁₉H₂₆O₁₃
MW: 462,40 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 104486-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47dc9ea23834332cc2ba25a7d62915177e7f95a8&vid=47dc9ea23834332cc2ba25a7d62915177e7f95a8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D +24.0 to +28.0 deg (C=1, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR