8-hydroxyquinoline-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 8-Hydroxyquinoline-2-carboxylic acid

H1658-1G H1658-5G
TCIAH1658-1GEA 70 EUR
TCIAH1658-1G TCIAH1658-5G
8-hydroxyquinoline-2-carboxylic acid ≥98.0% (by GC, titration analysis)
8-hydroxyquinoline-2-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189,17 g/mol
Número MDL: MFCD00168972
Núm. CAS: 1571-30-8

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR