Hexylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Hexan-1-amine hydrobromide, 1-Aminohexane hydrobromide, Hexylammonium bromide
H1678-1G
H1678-5G
TCIAH1678-1GEA
37
EUR
TCIAH1678-1G
TCIAH1678-5G
Hexylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Hexylamine hydrobromide
Formula:
C₆H₁₅N·HBr |
Núm. CAS:
7334-95-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Water | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...