Hydrocinchonidine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (8alpha,9R)-10,11-Dihydrocinchonan-9-ol, (R)-((1S,2S,4S,5R)-5-Ethylquinuclidin-2-yl)(quinolin-4-yl)methanol

H1702-1G H1702-5G
TCIAH1702-1GEA 94 EUR
TCIAH1702-1G TCIAH1702-5G
Hydrocinchonidine ≥98.0% (by HPLC, titration analysis)
Hydrocinchonidine
Formula: C₁₉H₂₄N₂O
MW: 296,41 g/mol
Número MDL: MFCD30489122
Núm. CAS: 485-64-3
EINECS: 207-620-2
Índice Merck: 14,04782

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 230.0 to 234.0 deg-C
Specific rotation [a]20/D -96.0 to -100.0 deg (C=0.3, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR