Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate ≥98.0% (por GC)
Proveedor: TCI
H1709-25G
H1709-5G
TCIAH1709-25GEA
70
EUR
TCIAH1709-25G
TCIAH1709-5G
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate ≥98.0% (por GC)
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate
Formula:
C₃₁H₅₄O₃ MW: 474,77 g/mol |
Número MDL:
MFCD00180757 Núm. CAS: 67845-93-6 EINECS: 267-342-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...