N-Hippuryl-L-histidyl-L-leucine hydrate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: N-Benzoyl-Gly-His-Leu, N-Benzoylglycyl-L-histidyl-L-leucine, N-Hippuryl-His-Leu

H1711-100MG H1711-25MG
TCIAH1711-100MGEA 124 EUR
TCIAH1711-100MG TCIAH1711-25MG
N-Hippuryl-L-histidyl-L-leucine hydrate ≥98.0% (by HPLC)
N-Hippuryl-L-histidyl-L-leucine hydrate
Formula: C₂₁H₂₇N₅O₅·xH₂O
MW: 429.48 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00149318
Núm. CAS: 207386-83-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.50 to 17.10 % (calcd.on anh.substance)
Specific rotation [a]20/D -40.0 to -44.0 deg (C=1, 1mol HCl)
Water 3.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR